missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAGE7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12945-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| GAGE7 | |
| Polyclonal | |
| Unconjugated | |
| GAGE7 | |
| AL4, Cancer/testis antigen 4.7, CT4.7, G antigen 7, GAGE 12I, GAGE 7, GAGE 7B, GAGE 8, GAGE family, GAGE12I, GAGE7, GAGE7B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2579 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O76087 | |
| GAGE7 | |
| GAGE7 Fusion Protein Ag4018 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction