missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAGE2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570663
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
GAGE2A is a protein coding gene.
Specifications
| GAGE2A | |
| Polyclonal | |
| Unconjugated | |
| GAGE2A | |
| cancer/testis antigen 4.2; cancer/testis antigen family 4, member 2; CT4.2; G antigen 2; G antigen 2A; G antigen 2A/2B; GAGE2; GAGE-2; GAGE2A; GAGE-2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 729447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6NT46 | |
| GAGE2A | |
| synthetic peptide directed towards the middle region of human GAGE2A (aa 66-115). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction