missing translation for 'onlineSavingsMsg'
Learn More

GAGE1 Antibody (4G6), Novus Biologicals™

Product Code. 18322218 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322218 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322218 Supplier Novus Biologicals Supplier No. H00002543M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAGE1 Monoclonal antibody specifically detects GAGE1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAGE1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4G6
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA, Sandwich ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001459.2
Gene Alias Antigen MZ2-F, Cancer/testis antigen 4.1, G antigen 1, GAGE-1, member 1, MGC33825, MZ2-F antigen
Host Species Mouse
Immunogen GAGE1 (NP_001459.2, 1 a.a. ~ 139 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQRQDPAAAQEGEDEGASAGQGPKPEADSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEEMRSHYVAQTGILWLLMNNCFLNLSPRKP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2543
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.