missing translation for 'onlineSavingsMsg'
Learn More

Gadoteric Acid, TRC

Product Code. 30390833
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390833 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390833 Supplier Toronto Research Chemicals Supplier No. TRCG12585525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gadoteric Acid
CAS 72573-82-1
Molecular Formula C16 H25 Gd N4 O8
Quantity 25 mg
Synonym Gadolinate(1-), [1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetato(4-)-κN1,κN4,κN7,κN10,κO1,κO4,κO7,κO10]-, hydrogen (1:1), Gadolinate(1-), [1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetato(4-)-N1,N4,N7,N10,O1,O4,O7,O10]-, hydrogen, Gadolinate(1-), [1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetato(4-)-κN1,κN4,κN7,κN10,κO1,κO4,κO7,κO10]-, hydrogen (9CI), 1,4,7,10-Tetraazacyclododecane, gadolinate(1-) deriv., 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, gadolinium complex, Gadoteric acid
InChI Formula InChI=1S/C16H28N4O8.Gd/c21-13(22)9-17-1-2-18(10-14(23)24)5-6-20(12-16(27)28)8-7-19(4-3-17)11-15(25)26;/h1-12H2,(H,21,22)(H,23,24)(H,25,26)(H,27,28);/q;+3/p-3
SMILES OC1=O[Gd+3]234567N8(CCN2(CCN3(CCN4(CC8)CC(=O)[O-]5)CC(=O)[O-]6)CC(=O)[O-]7)C1
Molecular Weight (g/mol) 558.64
Formula Weight 559.0913
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.