missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadopentetic acid, MedChemExpress
Description
The initial concentration of Gadopentetic acid (Gd-DTPA) in the plasma ([Gd-DTPA0]) is an important parameter for DCE-MRI. [Gd-DTPA0] is related to the administered bolus dose and varies with subjects. A bolus of 0.025 mmol/kg Gd-DTPA offers shorter acquisition time and less exposure of subjects, with a half-life of 37.3 mins, a mean residence time of 53.8 mins, and an AUC of 3.37 ± 0.47 mmol/min/L.
Specifications
Specifications
| Chemical Name or Material | Gadopentetic acid |
| Physical Form | Solid |
| CAS | 80529-93-7 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H20GdN3O10 |
| Synonym | Gd-DTPA gadolinium complex |
| Solubility Information | H2O : 100 mg/mL (182.63 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.83 mM; Need ultrasonic) |
| SMILES | O=C1[O-][Gd+3]23([O-]4)([O-]5)([O-]6)([O-]7)[N](CC[N]2(CC5=O)CC4=O)(CC[N]3(CC7=O)CC6=O)C1.[H+].[H+] |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?