missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium Standard: Gd @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17908619
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908619 500 mL 500mL
17939399 250 mL 250mL
17928439 50 mL 50mL
17989379 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17908619 Supplier VHG Labs Supplier No. VHGPGDN500
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Gadolinium Standard: Gd @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gadolinium
CAS 7440-54-2
Methods ICP/ICP-MS
Analyte or Component Names Gadolinium
Shipping Condition Room Temperature
Molecular Formula Gd
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Gd
SMILES [Gd]
IUPAC Name Gadolinium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 157
Quantity 500 mL
Formula Weight 157.25 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.