missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade

Product Code. 11480291
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480291 10 g 10g
11470291 2 g 2g
11490291 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480291 Supplier Thermo Scientific Alfa Aesar Supplier No. 040571.NC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14017-52-8

[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-52-8
MDL Number MFCD00150239
Synonym gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 563.69 (455.60 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Gadolinium(III) perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.