missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific™

Product Code. 10032500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032500 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10032500 Supplier Thermo Scientific Acros Supplier No. 202800500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 92°C
CAS 52788-53-1
Color White
Physical Form Granules
Quantity 50 g
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
Linear Formula Gd(NO3)3·5H2O
MDL Number MFCD00149729
Merck Index 14, 4326
Molecular Weight (g/mol) 433.33
Formula Weight 433.33
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.