missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate pentahydrate, 99.999%, (trace metal basis), ACROS Organics™

Product Code. 10502611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502611 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10502611 Supplier Thermo Scientific Acros Supplier No. 202800100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 92.0°C
CAS 52788-53-1
Color White
Physical Form Granules
Quantity 10 g
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
Linear Formula Gd(NO3)3·5H2O
MDL Number MFCD00149729
Merck Index 14, 4326
Formula Weight 433.33
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gadolinium(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.