missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO)

Product Code. 11331009
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331009 100 g 100g
11321009 5 g 5g
11341009 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331009 Supplier Thermo Scientific Alfa Aesar Supplier No. 011284.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94219-55-3 | GdN3O9 | 343.26 g/mol

Gadolinium(III) nitrate is used as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94219-55-3
Molecular Formula GdN3O9
Molecular Weight (g/mol) 343.26
MDL Number MFCD00011023
InChI Key MWFSXYMZCVAQCC-UHFFFAOYSA-N
IUPAC Name gadolinium(3+) trinitrate
SMILES [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 92°C
Color White
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Merck Index 14,4326
Solubility Information Soluble in water.
Formula Weight 343.26 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Density 2.406 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.