missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

£51.20 - £206.00

Chemical Identifiers

CAS 13765-26-9
Molecular Formula F3Gd
Molecular Weight (g/mol) 214.25
MDL Number MFCD00016113
InChI Key TYIZUJNEZNBXRS-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trifluoride
SMILES [F-].[F-].[F-].[Gd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331029
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011293.09
10 g
£51.20
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11321029
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011293.18
50 g
£206.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13765-26-9
214.25
TYIZUJNEZNBXRS-UHFFFAOYSA-K
[F-].[F-].[F-].[Gd+3]
F3Gd
MFCD00016113
gadolinium(3+) trifluoride

Specifications

1231°C
White
F3Gd
Insoluble in water.
[F-].[F-].[F-].[Gd+3]
214.25
99.99%
Odorless
7.1 g/mL
Gadolinium(III) fluoride, Anhydrous
13765-26-9
Crystalline
MFCD00016113
TYIZUJNEZNBXRS-UHFFFAOYSA-K
gadolinium(3+) trifluoride
214.25
Reacton™
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 237-369-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only