missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) chloride, ultra dry, 99.99% (REO)

Product Code. 11348607
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348607 25 g 25g
11358607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348607 Supplier Thermo Scientific Alfa Aesar Supplier No. 035736.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10138-52-0 | Cl3Gd | 263.6 g/mol

Gadolinium(III) Chloride is used as chelating agents, which is essential for biomedical applications. It is also employed in the synthesis and nuclear magnetic resonance (NMR) studies of paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes. It plays an important role as contrast agent for magnetic resonance imaging and an macrophage inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3Gd
Molecular Weight (g/mol) 263.6
MDL Number MFCD00011024
InChI Key MEANOSLIBWSCIT-UHFFFAOYSA-K
Synonym gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3
PubChem CID 61486
ChEBI CHEBI:37288
IUPAC Name trichlorogadolinium
SMILES Cl[Gd](Cl)Cl

Specifications

Melting Point 609°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 263.61
Sensitivity Hygroscopic
Appearance White powder
Chemical Name or Material Gadolinium(III) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.