missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11336658
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336658 5 g 5g
11346658 25 g 25g
11326658 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336658 Supplier Thermo Scientific Alfa Aesar Supplier No. 013208.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It acts as dopant material. It is employed in gamma-ray spectroscopy, medical imaging, nuclear and high energy physics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30010-20-9
Molecular Formula Br3Gd
Molecular Weight (g/mol) 396.96
MDL Number MFCD00049741
InChI Key KGOKDPWKDBWITQ-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Gd+3]

Specifications

Melting Point 186°C
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 396.96 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Gadolinium(III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.