missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
166.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It acts as dopant material. It is employed in gamma-ray spectroscopy, medical imaging, nuclear and high energy physics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 186°C |
Physical Form | Crystalline |
Quantity | 100 g |
Solubility Information | Soluble in water. |
Formula Weight | 396.96 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Gadolinium(III) bromide hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'