missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) acetate hydrate, 99.9% (REO)

Product Code. 11349399
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349399 25 g 25g
11339399 5 g 5g
11329399 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349399 Supplier Thermo Scientific Alfa Aesar Supplier No. 089158.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100587-93-7 | C6H9GdO6 | 334.38 g/mol

Gadolinium(III) acetate hydrate is used as a laboratory reagent, catalysts, optical glasses, structural ceramics, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-93-7
Molecular Formula C6H9GdO6
Molecular Weight (g/mol) 334.38
MDL Number MFCD00150116
InChI Key LYQGMALGKYWNIU-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) triacetate
SMILES [Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Boiling Point (decomposition)
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 334.39 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.611
Chemical Name or Material Gadolinium(III) acetate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Gadolinium(III) acetate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Gadolinium(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.