missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadodiamide, TRC
Specifications
Specifications
| Chemical Name or Material | Gadodiamide |
| CAS | 131410-48-5 |
| Molecular Formula | C16 H26 Gd N5 O8 |
| Quantity | 500 mg |
| Synonym | Gadolinium, [5,8-bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-κO)ethyl]-3-(oxo-κO)-2,5,8,11-tetraazatridecan-13-oato(3-)-κN5,κN8,κN11,κO13]-, Gadolinium, [5,8-bis(carboxymethyl)-11-[2-(methylamino)-2-oxoethyl]-3-oxo-2,5,8,11-tetraazatridecan-13-oato(3-)]-, 2,5,8,11-Tetraazatridecan-13-oic acid, 5,8-bis(carboxymethyl)-11-[2-(methylamino)-2-oxoethyl]-3-oxo-, gadolinium complex, [5,8-Bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-κO)ethyl]-3-(oxo-κO)-2,5,8,11-tetraazatridecan-13-oato(3-)-κN5,κN8,κN11,κO13]gadolinium, DV 7572, Gadodiamide, Gadolinium-DTPA-BMA, Gd DTPA-BMA, S 041, Gadolinium, [5,8-bis[(carboxy-kappaO)methyl]-11-[2-(methylamino)-2-(oxo-kappaO)ethyl]-3-(oxo-kappaO)-2,5,8,11-tetraazatridecan-13-oato(3-)-kappaN5,kappaN8,kappaN11,kappaO13]-, [5,8-Bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-kappaO)ethyl]-3-(oxo-kappaO)-2,5,8,11-tetraazatridecan-13-oato(3-)-kappaN5,kappaN8,kappaN11,kappaO13]gadolinium |
| InChI Formula | InChI=1S/C16H29N5O8.Gd/c1-17-12(22)7-20(10-15(26)27)5-3-19(9-14(24)25)4-6-21(11-16(28)29)8-13(23)18-2;/h3-11H2,1-2H3,(H,17,22)(H,18,23)(H,24,25)(H,26,27)(H,28,29);/q;+3/p-3 |
| SMILES | CNC1=O[Gd+3]234567N(CCN2(CC(=O)[O-]3)CC(=O4)NC)(CCN5(CC(=O)[O-]6)C1)CC(=O)[O-]7 |
| Molecular Weight (g/mol) | 573.66 |
| Formula Weight | 574.1022 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?