missing translation for 'onlineSavingsMsg'
Learn More

Gadodiamide, TRC

Product Code. 30400424
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400424 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400424 Supplier Toronto Research Chemicals Supplier No. TRCG12571050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gadodiamide
CAS 131410-48-5
Molecular Formula C16 H26 Gd N5 O8
Quantity 50 mg
Synonym Gadolinium, [5,8-bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-κO)ethyl]-3-(oxo-κO)-2,5,8,11-tetraazatridecan-13-oato(3-)-κN5,κN8,κN11,κO13]-, Gadolinium, [5,8-bis(carboxymethyl)-11-[2-(methylamino)-2-oxoethyl]-3-oxo-2,5,8,11-tetraazatridecan-13-oato(3-)]-, 2,5,8,11-Tetraazatridecan-13-oic acid, 5,8-bis(carboxymethyl)-11-[2-(methylamino)-2-oxoethyl]-3-oxo-, gadolinium complex, [5,8-Bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-κO)ethyl]-3-(oxo-κO)-2,5,8,11-tetraazatridecan-13-oato(3-)-κN5,κN8,κN11,κO13]gadolinium, DV 7572, Gadodiamide, Gadolinium-DTPA-BMA, Gd DTPA-BMA, S 041, Gadolinium, [5,8-bis[(carboxy-kappaO)methyl]-11-[2-(methylamino)-2-(oxo-kappaO)ethyl]-3-(oxo-kappaO)-2,5,8,11-tetraazatridecan-13-oato(3-)-kappaN5,kappaN8,kappaN11,kappaO13]-, [5,8-Bis[(carboxy-κO)methyl]-11-[2-(methylamino)-2-(oxo-kappaO)ethyl]-3-(oxo-kappaO)-2,5,8,11-tetraazatridecan-13-oato(3-)-kappaN5,kappaN8,kappaN11,kappaO13]gadolinium
InChI Formula InChI=1S/C16H29N5O8.Gd/c1-17-12(22)7-20(10-15(26)27)5-3-19(9-14(24)25)4-6-21(11-16(28)29)8-13(23)18-2;/h3-11H2,1-2H3,(H,17,22)(H,18,23)(H,24,25)(H,26,27)(H,28,29);/q;+3/p-3
SMILES CNC1=O[Gd+3]234567N(CCN2(CC(=O)[O-]3)CC(=O4)NC)(CCN5(CC(=O)[O-]6)C1)CC(=O)[O-]7
Molecular Weight (g/mol) 573.66
Formula Weight 574.1022
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.