missing translation for 'onlineSavingsMsg'
Learn More

Gadobutrol, MedChemExpress

Product Code. 16453057
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453057

Brand: Medchem Express HY16217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gadobutrol (Gd-DO3A-butrol; ZK 135079) is a nonionic paramagnetic macrocyclic gadolinium-based contrast agent that can be used for magnetic resonance imaging (MRI).

Gadobutrol (Gd-DO3A-butrol

Specifications

Chemical Name or Material Gadobutrol
Physical Form Solid
CAS 770691-21-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H31GdN4O9
Synonym ZK 135079
Solubility Information H2O : 20 mg/mL (33.07 mM; Need ultrasonic)
SMILES OC[C@H]1[N]23[Gd+3]([N]4(C5)CC3)([N]6(C7)CC4)([OH][C@@H]1CO)([O-]C5=O)([O-]C8=O)([O-]C7=O)[N](CC6)(C8)CC2
Molecular Weight (g/mol) 604.71
Formula Weight 604.71
Percent Purity 99.96%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.