missing translation for 'onlineSavingsMsg'
Learn More

gAdiponectin/gAcrp30 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18380102 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380102 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380102 Supplier Novus Biologicals Supplier No. NBP311025JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

gAdiponectin/gAcrp30 Polyclonal antibody specifically detects gAdiponectin/gAcrp30 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen gAdiponectin/gAcrp30
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Recombinant Human gAdiponectin/gAcrp30
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.