missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GADD45GIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550039
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total GADD45GIP1 protein.
Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
Specifications
| GADD45GIP1 | |
| Polyclonal | |
| Unconjugated | |
| Gadd45gip1 | |
| 2310040G17Rik; 39S ribosomal protein L59, mitochondrial; AI425883; CKBBP2; CKbetaBP2; CKII beta binding protein 2; CKII beta-associating protein; CR6 interacting factor 1; CR6-interacting factor 1; CRIF1; GADD45G interacting protein 1; GADD45GIP1; growth arrest- and DNA damage-inducible GADD45G-interacting protein; Growth arrest and DNA damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible, gamma interacting protein 1; Mitochondrial large ribosomal subunit protein mL64; Mrpl59; MRP-L59; p53-responsive gene 6 protein; papillomavirus L2 interacting nuclear protein 1; Papillomavirus L2-interacting nuclear protein 1; PLINP; PLINP1; PLINP-1; PRG6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90480 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TAE8 | |
| Gadd45gip1 | |
| Synthesized peptide derived from internal of human GADD45GIP1 (aa 91-140). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction