missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GADD45A Monoclonal Antibody (D.81.E)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA517014
This item is not returnable.
View return policy
Description
GADD45A Monoclonal Antibody for Western Blot
GADD45 alpha, beta and gamma are an evolutionarily conserved, homologous family of nuclear proteins that function as stress sensors for cellular physiological and environmental damage. GADD45 proteins are required for the activation of the G2/M checkpoint induced by UV radiation or alkylating agents. GADD45-induced G2/M checkpoint is regulated through inactivation of Cdc2-cyclin B1 kinase. GADD45 forms a complex with p21 and Cdc2, and may serve as core for interaction with other cell cycle regulators.
Specifications
| GADD45A | |
| Monoclonal | |
| 87 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P24522, P48316, P48317 | |
| GADD45A | |
| Synthetic peptide corresponding to amino acids surrounding Asn129 of human GADD45 alpha. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| D.81.E | |
| Unconjugated | |
| GADD45A | |
| AA545191; DDIT 1; DDIT1; DDIT-1; DNA damage-inducible transcript 1 protein; DNA damage-inducible transcript-1; DNA-damage inducible transcript 1; DNA-damage-inducible transcript 1; GADD45; GADD45A; Gadd45alpha; growth arrest and DNA damage inducible alpha; growth arrest and DNA damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible 45 alpha; growth arrest and DNA-damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible, alpha | |
| Rabbit | |
| Purified | |
| RUO | |
| 13197, 1647, 25112 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction