missing translation for 'onlineSavingsMsg'
Learn More

GADD153/CHOP Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18426684
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426684 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426684 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP213172AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GADD153/CHOP Polyclonal antibody specifically detects GADD153/CHOP in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GADD153/CHOP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C/EBP-homologous protein, C/EBP-homologous protein 10, CCAAT/enhancer-binding protein homologous protein, CHOP-10, CHOP10 CEBPZ, CHOPC/EBP zeta, DDIT-3, DNA damage-inducible transcript 3 protein, DNA-damage-inducible transcript 3, GADD153, Growth arrest and DNA damage-inducible protein GADD153
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human CHOP/GADD153 protein (between residues 100-150) [UniProt P35638]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, DNA Repair, Hypoxia, Lipid and Metabolism, Neuroscience, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 1649
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.