missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAD65 Monoclonal Antibody (ZG002)
Mouse Monoclonal Antibody
Brand: Invitrogen™ 398200
This item is not returnable.
View return policy
Description
GAD65 Monoclonal Antibody for ELISA, IHC (P), Western Blot
GAD65 (glutamic acid decarboxylase-65) are present in autoimmune disorders such as insulin-dependent (type1) diabetes mellitus (IDDM), stiff man syndrome and polyendocrine autoimmune disease. Auto-antibodies to GAD65 are present in 60-70% of individuals with newly diagnosed IDDM and thus are important markers for disease activity. These auto-antibodies usually recognize conformation-dependent regions on GAD65 and rarely bind to the second isoform, GAD67. Auto-antibodies to GAD67 are found in only 15% of recent-onset IDDM patients and most of this binding can be blocked with GAD65, suggesting shared epitopes between the two isoforms of GAD.
Specifications
| GAD65 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P48320, Q05329, Q05683 | |
| GAD2 | |
| Recombinant protein corresponding to the N-terminal region of the human GAD65 protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| ZG002 | |
| Unconjugated | |
| GAD2 | |
| 6330404F12Rik; 65 kDa glutamic acid decarboxylase; GAD(65); GAD2; Gad-2; GAD65; GAD-65; Glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase 2; glutamic acid decarboxylase 65; MGC161605; MGC161607; RP11-420F12.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 14417, 24380, 2572 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction