missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD2/GAD65 Mouse anti-Human, Clone: GAD2/2362, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | GAD2/GAD65 |
|---|---|
| Clone | GAD2/2362 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 μg/mL, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
GAD2/GAD65 Monoclonal specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| GAD2/GAD65 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 2572 | |
| IgG2b κ | |
| Protein A or G purified |
| GAD2/2362 | |
| Immunohistochemistry-Paraffin 0.1-0.2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
| Recombinant fragment of human GAD2/GAD65 protein (around aa 6-99) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title