missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD1/GAD67, Mouse anti-Human, Clone: GAD1/2563, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £396.00
Specifications
| Antigen | GAD1/GAD67 |
|---|---|
| Clone | GAD1/2563 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array |
| Applications | Western Blot, Flow Cytometry, ELISA, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18258686
|
Novus Biologicals
NBP2-79803-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18296416
|
Novus Biologicals
NBP2-79803-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array.Specifications
| GAD1/GAD67 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Diabetes Research | |
| 10 mM PBS with 0.05% BSA | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| GAD1 | |
| IgG2b κ | |
| Protein A or G purified | |
| This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GAD1/GAD67). |
| GAD1/2563 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q99259, Q99259 | |
| 2571 | |
| Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title