missing translation for 'onlineSavingsMsg'
Learn More

GAD1/GAD67 Mouse anti-Human, Clone: GAD1/2391, Novus Biologicals™

Product Code. 18371473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371473 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371473 Supplier Novus Biologicals Supplier No. NBP30735720UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAD1/GAD67
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone GAD1/2391
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Host Species Mouse
Immunogen Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2571
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.