missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD1/GAD67 Antibody (GAD1/2563) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79937
This item is not returnable.
View return policy
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array, CyTOF-ready.
Specifications
| GAD1/GAD67 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, ELISA, Protein Array, CyTOF-ready | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Peptide Array, CyTOF | |
| GAD1/2563 | |
| Unconjugated | |
| 10 mM PBS with No Preservative | |
| GAD1 | |
| Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) (Uniprot: Q99259) | |
| 100 μg | |
| Diabetes Research | |
| This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GDA1). There are two forms of glutamic acid decarboxylases (GADs) that are found in the brain: GAD65 (also known as GAD2) and GAD67 (also known as GAD1. GAD65 and GAD67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate-limiting step in the production of GABA (-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD65 localizes to synaptic vesicle membranes in nerve terminals, while GAD67 is distributed throughout the cell. GAD67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD65 will transiently activate to assist in GABA production. The loss of GAD65 is detrimental and can impair GABA neurotransmission, however the loss of GAD67 is lethal. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction