missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAD1/GAD67 Antibody (CL2911) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46637H
This item is not returnable.
View return policy
Description
GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GAD1/GAD67 | |
| Monoclonal | |
| HRP | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| This antibody was developed using a recombinant protein derived from Q99259, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Diabetes Research | |
| 2571 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL2911 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction