Learn More
Invitrogen™ GABRR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587640
Description
Immunogen sequence: TVQERKERKL REKFPCMCGM LHSKTMMLDG SYSESEANSL AGYPRSHILT EEERQDKIVV HLGLSGEANA ARKKGLLKGQ TGFRIFQNTH AIDKYSRLIF P; Positive Samples: HL-60, HepG2, Mouse heart; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA receptors, which are ligand-gated chloride channels. The protein encoded by this gene is a member of the rho subunit family and is a component of the GABA receptor complex.
Specifications
| GABRR2 | |
| Polyclonal | |
| Unconjugated | |
| Gabrr2 | |
| GABA(A) receptor; GABA(A) receptor subunit rho-2; GABA(C) receptor; GABA-C receptor, rho-2 subunit; Gabrr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 2; gamma-aminobutyric acid (GABA) C receptor, subunit rho 2; gamma-aminobutyric acid (GABA) receptor, rho 2; gamma-aminobutyric acid (GABA-A) receptor subunit rho 2; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 2; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 2; gamma-aminobutyric acid A receptor, rho 2; gamma-aminobutyric acid receptor rho2 subunit; gamma-aminobutyric acid receptor subunit rho-2; gamma-aminobutyric acid type A receptor rho2 subunit; gamma-aminobutyric acid type A receptor subunit rho2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14409, 2570 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P28476, P56476 | |
| Gabrr2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-450 of human GABRR2 (NP_0020343). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.