missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541008
This item is not returnable.
View return policy
Description
Peptide sequence: SNKSMTFDGR LVKKIWVPDV FFVHSKRSFT HDTTTDNIML RVFPDGHVLY Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA receptors, which are ligand-gated chloride channels. The protein encoded by this gene is a member of the rho subunit family and is a component of the GABA receptor complex.
Specifications
| GABRR2 | |
| Polyclonal | |
| Unconjugated | |
| Gabrr2 | |
| GABA(A) receptor; GABA(A) receptor subunit rho-2; GABA(C) receptor; GABA-C receptor, rho-2 subunit; Gabrr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 2; gamma-aminobutyric acid (GABA) C receptor, subunit rho 2; gamma-aminobutyric acid (GABA) receptor, rho 2; gamma-aminobutyric acid (GABA-A) receptor subunit rho 2; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 2; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 2; gamma-aminobutyric acid A receptor, rho 2; gamma-aminobutyric acid receptor rho2 subunit; gamma-aminobutyric acid receptor subunit rho-2; gamma-aminobutyric acid type A receptor rho2 subunit; gamma-aminobutyric acid type A receptor subunit rho2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2570 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P28476 | |
| Gabrr2 | |
| Synthetic peptide directed towards the middle region of human GABRR2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction