missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRR2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577406
This item is not returnable.
View return policy
Beskrivning
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA receptors, which are ligand-gated chloride channels. The protein encoded by this gene is a member of the rho subunit family and is a component of the GABA receptor complex.
Specifikationer
| GABRR2 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Gabrr2 | |
| Gabrr2, GABA(C) receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29695 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P47742 | |
| Gabrr2 | |
| Peptide (C)RKRWTGHLETSKPSH, corresponding to amino acid residues 51-65 of rat GABA(A) rho2 | |
| 50 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering