Learn More
Invitrogen™ GABRQ Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589095
Description
Immunogen sequence: NYLFYSRGPR RQPRRHRRPR RVIARYRYQQ VVVGNVQDGL INVEDGVSSL PITPAQAPLA SPESLGSLTS TSEQAQLATS ESLSPLTSLS GQAPLATGES LSDLPSTSEQ ARHSYGVRFN GFQADDSIIP TEIRNRVEAH GHGVTHDHED SNESLSSDER HGHGPSGKPM LHHGEKGVQE AGWDLDDNND KSDCLAIKEQ FKCDTNSTWG LNDDELMAHG QEKDSSSESE DSCPPSPGCS FTEGFSFDLF N; Positive Samples: Jurkat; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. GABRQ gene is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases:early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. It is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases: early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).
Specifications
| GABRQ | |
| Polyclonal | |
| Unconjugated | |
| GABRQ | |
| GABA(A) receptor subunit theta; GABA(A) Theta; GABRQ; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid (GABA) A receptor, subunit theta; gamma-aminobutyric acid (GABA) A receptor, theta; gamma-aminobutyric acid (GABA) receptor, theta; gamma-aminobutyric acid (GABA-A) receptor, subunit theta; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid receptor subunit theta; gamma-aminobutyric acid type A receptor subunit theta; gamma-aminobutyric acid type A receptor theta subunit; THETA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UN88 | |
| GABRQ | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-600 of human GABRQ (NP_0610283). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.