missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111385
This item is not returnable.
View return policy
Description
Immunogen sequence: ARSRKVEEDE YEDSSSNQKW VLAPKSQDTD VTLILNKLLR EYD Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 93%.
GABRG3 is a family of proteins involved in the GABAergic neurotransmission of the mammalian central nervous system. GABRG3 is a member of the GABA-A receptor gene family of heteromeric pentameric ligand-gated ion channels through which GABA, the major inhibitory neurotransmitter in the mammalian brain, acts. GABA-A receptors are the site of action of a number of important pharmacologic agents including barbiturates, benzodiazepines, and ethanol (summary by Whiting et al., 1999). For additional general information about the GABA-A receptor gene family, see GABRA1 (MIM 137160).
Specifications
| GABRG3 | |
| Polyclonal | |
| Unconjugated | |
| GABRG3 | |
| B230362M20Rik; CAE2; ECA2; GABA(A) receptor subunit gamma-3; GABA(G) receptor; GABA(G) receptor, gamma 3; GABA-alpha receptor gamma-3 subunit; GABRG3; Gabrg-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, gamma 3; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 3; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 3; gamma-aminobutyric acid A receptor, gamma 3; gamma-aminobutyric acid receptor subunit gamma-3; gamma-aminobutyric acid type A receptor gamma3 subunit; GEFSP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2567 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99928 | |
| GABRG3 | |
| Recombinant Protein corresonding to Human GABRG3. Recombinant protein control fragment (Product #RP-101533). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction