missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12871-1-AP-20UL
This item is not returnable.
View return policy
Description
Gabrg1 encodes a protein that belongs to the ligand-gated ionic channel family. Gabrg1 is an integral membrane protein and plays an important role in inhibiting neurotransmission by binding to the benzodiazepine receptor and opening an integral chloride channel. The Gabrg1 gene is clustered with three other family members on chromosome 4. GABAA receptors are members of the Cys-loop family of ligand-gated ion channels and, along with GABAB receptors, are responsible for mediating the inhibitory effects of GABA. Structurally, Gabrg1 are pentameric proteins, consisting of five subunits belonging to different families. Diseases associated with GABRG1 include Alcohol Dependence.Specifications
| GABRG1 | |
| Polyclonal | |
| Unconjugated | |
| GABRG1 | |
| GABA A receptor gamma 1, GABAA receptor gamma 1, GABRG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140674, 14405, 2565 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23574, Q8N1C3, Q9R0Y8 | |
| Gabrg1 | |
| GABRG1 Fusion Protein Ag3535 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction