missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583722
This item is not returnable.
View return policy
Description
Immunogen sequence: KVKVSRPRAE MDVRNAIVLF SLSAAGVTQE LAISRRQRRV PGNLMGSYRS VGVETGETKK EGAARSGGQG GIRARL Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 82%.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
Specifications
| GABRD | |
| Polyclonal | |
| Unconjugated | |
| GABRD | |
| AI853201; EIG10; EJM7; GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABAA receptor delta subunit; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA-RD; GABAARdelta; Gabrd; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, subunit delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid receptor subunit delta; gamma-aminobutyric acid type A receptor delta subunit; GEFSP5; MGC105467; MGC45284 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2563 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14764 | |
| GABRD | |
| Recombinant protein corresponding to Human GABRD. Recombinant protein control fragment (Product #RP-88808). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction