Learn More
Invitrogen™ GABRB2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA595972
Product Code. 16568811
  • £440.00 / 100µL
Description
Immunogen sequence: SVNDPSNMSL VKETVDRLLK GYDIRLRPDF GGPPVAVGMN IDIASIDMVS EVNMDYTLTM YFQQAWRDKR LSYNVIPLNL TLDNRVADQL WVPDTYFLND KKSFVHGVTV KNRMIRLHPD GTVLYGLRIT TTAACMMDLR RYPLDEQNCT LEIESYGYTT DDIEFYWRGD DNAVTGVTKI ELPQFSIVDY KLITKKVVFS TGSYPRLSLS FKLKRNIGY; Positive Samples: Mouse liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA Rα1-6, GABAA R β1-3, GABAA Rγ1-3, GABAA Rδ, GABAA Rε, GABAA Rρ1 and GABAA Rρ2. The GABAB family is composed of GABAB R1α and GABAB R1β. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GABRB2
Polyclonal
Unconjugated
GABRB2
AI834970; C030002O17Rik; C030021G16Rik; GABA(A) receptor subunit beta-2; GABAARbeta2; Gabrab2; GABRB2; Gabrb-2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 2; gamma-aminobutyric acid (GABA) A receptor, subunit beta 2; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 2; gamma-aminobutyric acid A receptor beta 2; gamma-aminobutyric acid A receptor, subunit beta 2; Gamma-aminobutyric acid receptor beta 2; gamma-aminobutyric acid receptor beta-2 subunit; gamma-aminobutyric acid receptor subunit beta-2; gamma-aminobutyric acid receptor, subunit beta 2; gamma-aminobutyric acid type A receptor beta2 subunit; GARB2; MGC119386; MGC119388; MGC119389
Rabbit
Affinity chromatography
RUO
14401, 2561
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P47870, P63137
GABRB2
Recombinant fusion protein containing a sequence corresponding to amino acids 26-244 of human GABA A Receptor beta 2 (GABA A Receptor beta 2 (GABRB2)) (NP_0008041).
100 μL
Primary
Human, Mouse