missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595657
This item is not returnable.
View return policy
Description
Antigen Affinity Purified from Pooled Serum.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
Specifications
| GABRB1 | |
| Polyclonal | |
| Unconjugated | |
| Gabrb1 | |
| AW061132; B230208N19Rik; GABA beta subunit; GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; Gabrb1; Gabrb-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, subunit beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; gamma-aminobutyric acid type A receptor beta1 subunit; gamma-aminobutyric acid type A receptor subunit beta1; GARB1; GBRB1; si:dkey-20i10.4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14400, 25450 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P15431, P50571 | |
| Gabrb1 | |
| Fusion protein from the cytoplasmic loop of the beta 1 subunit. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction