missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567443
This item is not returnable.
View return policy
Description
Immunogen sequence: FGFSQMPTSSV KDETNDNITI FT Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
GABRA5 is a part of GABA, a major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described.
Specifications
| GABRA5 | |
| Polyclonal | |
| Unconjugated | |
| GABRA5 | |
| A230018I05Rik; GABA(A) receptor; GABA(A) receptor subunit alpha-5; GABAA receptor alpha5 subunit; GABA-A receptor alpha-5 subunit; GABRA5; Gabra-5; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 5; gamma-aminobutyric acid (GABA) A receptor, alpha 5; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 5; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 5; gamma-aminobutyric acid A receptor, alpha 5; gamma-aminobutyric acid receptor subunit alpha-5; gamma-aminobutyric acid type A receptor alpha5 subunit; ligand-gated ion channel subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31644 | |
| GABRA5 | |
| Recombinant Human GABRA5. Recombinant protein control fragment (Product #RP-108105). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction