missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12979-1-AP-150UL
This item is not returnable.
View return policy
Description
Gamma-aminobutyric acid is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. This gene encodes subunit alpha-4, which is involved in the etiology of autism and eventually increases autism risk through interaction with another subunit, gamma-aminobutyric acid receptor beta-1. Alternatively spliced transcript variants encoding different isoforms have been found in this gene.Specifications
| GABRA4 | |
| Polyclonal | |
| Unconjugated | |
| GABRA4 | |
| GABA A Receptor alpha 4, GABAA Receptor alpha 4, GABRA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140675, 14397, 2557 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28471, P48169, Q9D6F4 | |
| Gabra4 | |
| GABRA4 Fusion Protein Ag3600 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction