Learn More
GABRA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12979-1-AP-20UL
 View more versions of this product
Product Code. 16890964
  • £135.00 / 20µL
Description
Gamma-aminobutyric acid is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. This gene encodes subunit alpha-4, which is involved in the etiology of autism and eventually increases autism risk through interaction with another subunit, gamma-aminobutyric acid receptor beta-1. Alternatively spliced transcript variants encoding different isoforms have been found in this gene.
Specifications
Specifications
GABRA4
Polyclonal
Unconjugated
GABRA4
GABA A Receptor alpha 4, GABAA Receptor alpha 4, GABRA4
Rabbit
Antigen Affinity Chromatography
RUO
140675, 14397, 2557
-20°C
Liquid
Immunoprecipitation, Western Blot
0.57 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28471, P48169, Q9D6F4
Gabra4
GABRA4 Fusion Protein Ag3600
20 μL
Primary
Mouse, Rat, Human