Learn More
GABRA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540998
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AEVVYSWTLG KNKSVEVAQD GSRLNQYDLL GHVVGTEIIR SSTGEYVVMT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
GABRA3 Polyclonal antibody specifically detects GABRA3 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GABRA3 | |
Polyclonal | |
Unconjugated | |
Gabra3 | |
GABA(A) receptor; GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABA-A receptor alpha-3 subunit; Gabra3; Gabra-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 3; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 3; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 3; gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid receptor, subunit alpha 3; gamma-aminobutyric acid type A receptor alpha3 subunit; gamma-aminobutyric acid type A receptor subunit alpha3; Gamma-animobutyric acid (GABA) A receptor alpha 3; gamma-animobutyric acid (GABA) A receptor, alpha 3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid type A receptor subunit alpha3; MGC33793; unnamed protein product | |
Rabbit | |
Protein A | |
RUO | |
14396, 2556 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P26049, P34903 | |
Gabra3 | |
Synthetic peptide directed towards the middle region of human GABRA3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |