missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABPB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569724
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
GABPB2 (GA binding protein transcription factor, beta subunit 2) is a protein-coding gene. GO annotations related to this gene include protein homodimerization activity and transcription regulatory region DNA binding. An important paralog of this gene is ANKRD2.
Specifications
| GABPB2 | |
| Polyclonal | |
| Unconjugated | |
| Gabpb2 | |
| 1810015F01Rik; 5830427M07; 9430006E19Rik; A430024B14Rik; AV050852; BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor beta subunit 2; GA binding protein transcription factor subunit beta 2; GA binding protein transcription factor, alpha subunit pseudogene; GA binding protein transcription factor, beta subunit 1; GA binding protein transcription factor, beta subunit 2; GA repeat binding protein, beta 2; GA-binding protein beta-2-1; GA-binding protein subunit beta-1; GA-binding protein subunit beta-2; GABP subunit beta-1; GABP subunit beta-2; GABP subunit beta-2-1; GABP2; GABPAP; GABPB; GABPB1; GABPB-1; Gabpb2; GABPB-2; Gabpb2-1; LOW QUALITY PROTEIN: GA-binding protein subunit beta-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1565234; RP11-68I18.1; Transcription factor E4TF1-47; transcription factor E4TF1-53 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 295263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Gabpb2 | |
| synthetic peptide directed towards the N-terminal region of rat GABPB2. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction