missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabpb2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38042-20ul
This item is not returnable.
View return policy
Description
Gabpb2 Polyclonal antibody specifically detects Gabpb2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Gabpb2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| GA binding protein transcription factor, beta subunit 2, GA-binding protein subunit beta-2, GABP subunit beta-2, GABPB-2, MGC29891 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 239-448 of human Gabpb2 (NP_653219.1).,, Sequence:, SNSHRATANTEEIIEGNSVDSSIQQVMGSGGQRVITIVTDGVPLGNIQTSIPTGGIGQPFIVTVQDGQQVLTVPAGKVAEETVIKEEEEEKLPLTKKPRIGEKTNSVEESKEGNERELLQQQLQEANRRAQEYRHQLLKKEQEAEQYRLKLEAIARQQPNGVDFTMVEEVAEVDAVVVTEGELEERETKVTGSAGTTEPHTRVSMATVSS | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 126626 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction