missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABPB1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00002553-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MSLVDLGKKLLEAARAGQDDEVRILMANGAPFTTDWLGTSPLHLAAQYGHYSTTEVLLRAGVSRDARTKVDRTPLHMAASEGHASIVEVLLKHGADVNAKDMLKMTALHWATEHNHQEVVELLIKYGADVHTQSKFCKTAFDISIDNGNEDLAEILQIAMQNQINTNPESPDTVTIHAATPQFIIGPGGVVNLTGLVSSENSSKATDETGVSAVQFGNSSTSVLATLAALAEASAPLSNSSETPVVATEEVVTAESVDGAIQQVVSSGGQQVITIVTDGIQLGNLHSIPTSGIGQPIIVTMPDGQQVLTVPATDIAEETVISEEPPAKRQCIEIIENRVESAEIEEREALQKQLDEANREAQKYRQQLLKKEQEAEAYRQKLEAMTRLQTNKEAVSpecifications
| GABPB1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_005254.4 | |
| GABPB1 | |
| GABPB1 (NP_005245.2, 1 a.a. ∼ 395 a.a) full-length human protein. | |
| RUO | |
| 2553 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| GA binding protein transcription factor, beta subunit 1 | |
| BABPB2/E4TF1/E4TF1-47/E4TF1-53/E4TF1B/GABPB/GABPB2/NRF2B1/NRF2B2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction