missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABPB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115263
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GABPB1.
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
Specifications
| GABPB1 | |
| Polyclonal | |
| Unconjugated | |
| GABPB1 | |
| BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4Tf1B; GA binding protein subunit beta1; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA repeat binding protein, beta 1; GA-binding protein subunit beta-1; Gabp beta1; GABP subunit beta-1; GABP subunit beta-2; GABPB; Gabpb1; GABPB-1; GABPB1-1; GABPB1-2; GABPB2; GABPB-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1560391; transcription factor E4TF1-47; Transcription factor E4TF1-53 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14391, 2553, 499883 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q00420, Q06547 | |
| GABPB1 | |
| A synthesized peptide derived from human GABPB1(Accession Q06547), corresponding to amino acid residues V264-D314. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction