missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABPB1 Monoclonal Antibody (OTI7A7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527434
This item is not returnable.
View return policy
Description
GABPB1 Monoclonal Antibody for Western Blot, IHC (P)
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
Specifications
| GABPB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q06547 | |
| GABPB1 | |
| Human recombinant protein fragment corresponding to amino acids 128-360 of GABPB1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7A7 | |
| Unconjugated | |
| GABPB1 | |
| BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4Tf1B; GA binding protein subunit beta1; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA repeat binding protein, beta 1; GA-binding protein subunit beta-1; Gabp beta1; GABP subunit beta-1; GABP subunit beta-2; GABPB; Gabpb1; GABPB-1; GABPB1-1; GABPB1-2; GABPB2; GABPB-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1560391; transcription factor E4TF1-47; Transcription factor E4TF1-53 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2553 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction