missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588319
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, Raji, HepG2, SKOV3, Rat liver; Cellular Location: Nucleus Immunogen sequence: NLKKLLEPRL QCSLDAHEIC LQDIQLDPER SLFDQGVKTD GTVQLSVQVI SYQGIEPKLN ILEIVKPADT VEVVIDPDAH HAESEAHLVE EAQVITLDGT K.
This gene encodes one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| GABPA | |
| Polyclonal | |
| Unconjugated | |
| GABPA | |
| E4TF1-60; E4TF1A; GA binding protein transcription factor alpha subunit; GA binding protein transcription factor alpha subunit 60kDa; GA binding protein transcription factor, alpha subunit; GA binding protein transcription factor, alpha subunit 60kDa; GA repeat binding protein alpha; GA repeat binding protein, alpha; GA-binding protein alpha chain; GA-binding protein alpha-subunit; GABP subunit alpha; GABPA; GABPalpha; human nuclear respiratory factor-2 subunit alpha; NFT2; NRF2; Nrf2a; Nrf-2alpha; nuclear respiratory factor 2 alpha; nuclear respiratory factor 2 alpha subunit; nuclear respiratory factor 2 subunit alpha; RCH04A07; Transcription factor E4TF1-60 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2551, 363735 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q06546 | |
| GABPA | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human GABPA (NP_0020312). | |
| 100 μL | |
| Primary | |
| Human, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction