Learn More
Memmert™ Gable Cover for Shaking Device/Peltier Cooling Unit
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
14/22 and 29/45 Shaking device, CDP 115 Peltier cooling unit Model 7
Unit Size:
Each
Description
- Stainless steel construction
- Durable
- Corrosion resistant
- Easy to clean
- Perfectly fitted
Specifications
Specifications
| For Use With (Equipment) | 14/22 and 29/45 Shaking device, CDP 115 Peltier cooling unit Model 7 |
| Material | Stainless Steel |
| Warranty | 3 Years |