missing translation for 'onlineSavingsMsg'
Learn More

Gabexate mesylate, MedChemExpress

Product Code. 16458468
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458468 10 mg 10mg
16458276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458468 Supplier Medchem Express Supplier No. HYB0385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gabexate mesylate is a Factor X inhibitor; serine protease inhibitor .

Gabexate mesylate is a Factor X inhibitor

Specifications

Chemical Name or Material Gabexate mesylate
CAS 56974-61-9
Color White
Purity Grade Notes Research
Molecular Formula C17H27N3O7S
Quantity 10 mg
Synonym FOY
Solubility Information DMSO : ≥ 100 mg/mL (239.53 mM) ∣H2O : 7.14 mg/mL (17.10 mM; Need ultrasonic)
SMILES O=C(OCC)C1=CC=C(OC(CCCCCNC(N)=N)=O)C=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 417.48
Formula Weight 417.48
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.