Learn More
Invitrogen™ GABBR2 Recombinant Rabbit Monoclonal Antibody (19H3L12)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703357
Beschreibung
This antibody is predicted to react with Monkey, Mouse, Dog. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The multi-pass membrane protein, GABBR2, belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Spezifikation
| GABBR2 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75899, O88871 | |
| GABBR2 | |
| Peptide corresponding to human GABBR2 [ aa318-aa333]. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| 19H3L12 | |
| Unconjugated | |
| GABBR2 | |
| FLJ36928; G protein-coupled receptor 51; GABA BR; GABA-B R2 receptor; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABABR2; GABA-BR2; GABA-B-R2; Gabbr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; Gm425; Gpr51; GPRC3B; G-protein coupled receptor 51; HG20; HRIHFB2099; MGC119386; MGC119388; MGC119389; ortholog of human G protein-coupled receptor 51 GPR51; RP23-337N2.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 83633, 9568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.